siRNA Details
| Gene Symbol: Dbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031024.1 | NP_112286.1 | 10, 11 | 1015 | ||
| XM_006253655.2 | XP_006253717.1 | 10, 11 | 1101 | |||
| GenBank mRNA | X59267.1 | - | 10, 11 | 1015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbn1| Gene Symbol: Dbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031024.1 | NP_112286.1 | 10, 11 | 1015 | ||
| XM_006253655.2 | XP_006253717.1 | 10, 11 | 1101 | |||
| GenBank mRNA | X59267.1 | - | 10, 11 | 1015 | ||