siRNA Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031025.1 | NP_112287.1 | 13 | 1656 | ||
| GenBank mRNA | BC107440.1 | - | 13 | 1656 | 7115402 | |
| D00092.1 | - | 10 | 1160 | |||
| D10655.1 | - | 12 | 1373 | |||
| M16075.1 | - | 10 | 1160 | |||

Dlat
Genomic Map