siRNA Details
| Gene Symbol: Dync1li2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031026.1 | NP_112288.1 | 12 | 1473 | ||
| XM_006255053.2 | XP_006255115.1 | 13 | 1672 | |||
| XM_006255054.2 | XP_006255116.1 | 12 | 1630 | |||
| GenBank mRNA | AB008521.1 | - | 8 | 724 | ||
| AB008522.1 | - | 7 | 739 | |||
| AB008523.1 | - | 7 | 682 | |||
| AB008524.1 | - | 8 | 781 | |||
| BC089794.1 | - | 13 | 1479 | 7115029 | ||
| U15138.1 | - | 12 | 1473 | |||

Dync1li2
Genomic Map