siRNA Details
Gene Symbol: Gatm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031031.2 | NP_112293.1 | 2 | 184 | ||
GenBank mRNA | BC081785.1 | - | 2 | 184 | 7132560 | |
U07971.1 | - | 2 | 176 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gatm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031031.2 | NP_112293.1 | 2 | 184 | ||
GenBank mRNA | BC081785.1 | - | 2 | 184 | 7132560 | |
U07971.1 | - | 2 | 176 |