siRNA Details
| Gene Symbol: Gnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031037.2 | NP_112299.1 | 8 | 1005 | ||
| GenBank mRNA | AF277892.1 | - | 7 | 786 | ||
| AF397193.1 | - | 4 | 585 | |||
| BC065579.1 | - | 8 | 1005 | 5598923 | ||
| DQ120485.1 | - | 7 | 770 | |||
| DQ120486.1 | - | 7 | 770 | |||
| FQ209560.1 | - | 4 | 393 | |||
| U34959.1 | - | 7 | 777 | |||

Gnb2
Genomic Map