siRNA Details
| Gene Symbol: Gpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031039.1 | NP_112301.1 | 3, 4 | 475 | ||
| XM_008765599.1 | XP_008763821.1 | 4, 5 | 1195 | |||
| GenBank mRNA | BC085728.1 | - | 2, 3 | 530 | ||
| BC097937.1 | - | 3, 4 | 479 | 7324977 | ||
| D10354.1 | - | 3, 4 | 475 | |||

Gpt
Genomic Map