siRNA Details
Gene Symbol: Gtf2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031042.3 | NP_112304.2 | 3, 4 | 267 | ||
GenBank mRNA | BC091112.1 | - | 3, 4 | 267 | 7381243 | |
D10665.1 | - | 3, 4 | 317 | |||
FQ232071.1 | - | 3, 4 | 306 | |||
FQ233324.1 | - | 3, 4 | 306 | |||
L01267.1 | - | 3, 4 | 244 |
