siRNA Details
| Gene Symbol: Gtf2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031042.3 | NP_112304.2 | 6 | 558 | ||
| GenBank mRNA | BC091112.1 | - | 6 | 558 | 7381243 | |
| D10665.1 | - | 6 | 608 | |||
| FQ232071.1 | - | 6 | 597 | |||
| FQ233324.1 | - | 6 | 597 | |||
| L01267.1 | - | 6 | 535 | |||

Gtf2f2
Genomic Map