siRNA Details
| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031057.2 | NP_112319.2 | 1, 2 | 93 | ||
| GenBank mRNA | CB760062.1 | - | 1, 2 | 150 | ||
| DN948946.1 | - | 1, 2 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh6a1| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031057.2 | NP_112319.2 | 1, 2 | 93 | ||
| GenBank mRNA | CB760062.1 | - | 1, 2 | 150 | ||
| DN948946.1 | - | 1, 2 | 90 | |||