siRNA Details
| Gene Symbol: Nell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031070.2 | NP_112332.2 | 6 | 607 | ||
| XM_008765797.1 | XP_008764019.1 | 5 | 957 | |||
| XM_008765798.1 | XP_008764020.1 | 6 | 578 | |||
| GenBank mRNA | AY089719.1 | - | 6 | 607 | ||
| BC093617.1 | - | 5 | 551 | 7312010 | ||
| EF110908.1 | - | 4 | 400 | |||
| GQ376510.1 | - | 5 | 529 | |||
| U48245.1 | - | 6 | 607 | |||

Nell2
Genomic Map