siRNA Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004132.1 | NP_001004132.1 | 11 | 1140 | ||
| NM_031077.1 | NP_112339.1 | 11 | 1499 | |||
| XM_006256617.2 | XP_006256679.1 | 11 | 5267 | |||
| XM_006256618.2 | XP_006256680.1 | 11 | 1500 | |||
| XM_006256619.2 | XP_006256681.1 | 11 | 1552 | |||
| XM_008773037.1 | XP_008771259.1 | 11 | 5270 | |||
| GenBank mRNA | BC078711.1 | - | 11 | 1140 | 7112874 | |
| U36444.1 | - | 11 | 1499 | |||

Cdk16
Genomic Map