siRNA Details
| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031080.1 | NP_112342.1 | 2 | 300 | ||
| XM_006232188.2 | XP_006232250.1 | 2 | 991 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde7a| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031080.1 | NP_112342.1 | 2 | 300 | ||
| XM_006232188.2 | XP_006232250.1 | 2 | 991 | |||