siRNA Details
| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031080.1 | NP_112342.1 | 4 | 513 | ||
| XM_006232188.2 | XP_006232250.1 | 4 | 1204 | |||
| XM_008760880.1 | XP_008759102.1 | 3 | 1339 | |||
| GenBank mRNA | U77880.1 | - | 3 | 218 | ||

Pde7a
Genomic Map