siRNA Details
| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031085.2 | NP_112347.1 | 1 | 368 | ||
| GenBank mRNA | BC061763.1 | - | 1 | 543 | ||
| BC081782.1 | - | 1 | 368 | 7123785 | ||
| X68400.1 | - | 1 | 209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkch| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031085.2 | NP_112347.1 | 1 | 368 | ||
| GenBank mRNA | BC061763.1 | - | 1 | 543 | ||
| BC081782.1 | - | 1 | 368 | 7123785 | ||
| X68400.1 | - | 1 | 209 | |||