siRNA Details
| Gene Symbol: Rps6ka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031107.1 | NP_112369.1 | 4 | 399 | ||
| XM_006239083.2 | XP_006239145.1 | 3 | 803 | |||
| XM_006239084.2 | XP_006239146.1 | 3 | 719 | |||
| GenBank mRNA | M99169.1 | - | 4 | 399 | ||

Rps6ka1
Genomic Map