siRNA Details
| Gene Symbol: Ccl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031116.3 | NP_112378.2 | 2, 3 | 227 | ||
| GenBank mRNA | BC058138.1 | - | 2, 3 | 243 | 6921268 | |
| EF121972.1 | - | 185 | ||||
| EF122007.1 | - | 2, 3 | 185 | |||
| U06436.1 | - | 2, 3 | 185 | |||

Ccl5
Genomic Map