siRNA Details
| Gene Symbol: Ccl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031116.3 | NP_112378.2 | 1 | 63 | ||
| GenBank mRNA | BC058138.1 | - | 1 | 79 | 6921268 | |
| EF121972.1 | - | 21 | ||||
| EF122007.1 | - | 1 | 21 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl5| Gene Symbol: Ccl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031116.3 | NP_112378.2 | 1 | 63 | ||
| GenBank mRNA | BC058138.1 | - | 1 | 79 | 6921268 | |
| EF121972.1 | - | 21 | ||||
| EF122007.1 | - | 1 | 21 | |||