siRNA Details
Gene Symbol: Nr2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031130.1 | NP_112392.1 | 2 | 1365 | ||
GenBank mRNA | FQ212534.1 | - | 1 | 225 | ||
U10995.1 | - | 2 | 1365 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031130.1 | NP_112392.1 | 2 | 1365 | ||
GenBank mRNA | FQ212534.1 | - | 1 | 225 | ||
U10995.1 | - | 2 | 1365 |