siRNA Details
| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031132.3 | NP_112394.3 | 3 | 685 | ||
| XM_008766690.1 | XP_008764912.1 | 4 | 1241 | |||
| GenBank mRNA | GU067683.1 | - | 3 | 434 | ||
| L09653.1 | - | 3 | 492 | |||
| S67770.1 | - | 3 | 685 | |||

Tgfbr2
Genomic Map