siRNA Details
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031138.2 | NP_112400.1 | 2, 3 | 234 | ||
XM_006246343.2 | XP_006246405.1 | 3, 4 | 259 | |||
GenBank mRNA | BC070946.1 | - | 2, 3 | 227 | 7103580 | |
DN931313.1 | - | 2, 3 | 264 | |||
FQ216310.1 | - | 1 | 14 | |||
FQ222428.1 | - | 2, 3 | 251 | |||
FQ229894.1 | - | 2, 3 | 245 | |||
M62388.1 | - | 2, 3 | 176 |
Gene Symbol: LOC103694902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773893.1 | XP_008772115.1 | 3, 4 | 258 |
