siRNA Details
| Gene Symbol: Pax8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031141.2 | NP_112403.2 | 10 | 1348 | ||
| XM_006233646.2 | XP_006233708.1 | 10 | 2448 | |||
| XM_008761610.1 | XP_008759832.1 | 9 | 2784 | |||
| XM_008761611.1 | XP_008759833.1 | 8 | 2578 | |||
| XM_008761612.1 | XP_008759834.1 | 7 | 2512 | |||
| XM_008761613.1 | XP_008759835.1 | 8 | 2573 | |||
| GenBank mRNA | BC081779.1 | - | 10 | 1348 | 7127710 | |
| FQ221890.1 | - | 2 | 197 | |||
| X94246.1 | - | 9 | 1231 | |||

Pax8
Genomic Map