siRNA Details
| Gene Symbol: Cds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031242.2 | NP_112521.2 | 5, 6 | 629 | ||
| GenBank mRNA | AB009999.1 | - | 574 | |||
| BC127492.1 | - | 5, 6 | 629 | 8373547 | ||
| FQ212493.1 | - | 604 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cds1| Gene Symbol: Cds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031242.2 | NP_112521.2 | 5, 6 | 629 | ||
| GenBank mRNA | AB009999.1 | - | 574 | |||
| BC127492.1 | - | 5, 6 | 629 | 8373547 | ||
| FQ212493.1 | - | 604 | ||||