siRNA Details
| Gene Symbol: Dynlt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031318.1 | NP_112608.1 | 3, 4 | 196 | ||
| XM_006227901.2 | XP_006227963.1 | 3, 4 | 261 | |||
| GenBank mRNA | AB010119.1 | - | 3, 4 | 185 | ||
| AJ131437.1 | - | 3, 4 | 196 | |||
| BC166879.1 | - | 3, 4 | 228 | 6919674 | ||

Dynlt1
Genomic Map