siRNA Details
| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031323.1 | NP_112613.1 | 5, 6 | 871 | ||
| XM_006252285.1 | XP_006252347.1 | 2, 3 | 344 | |||
| XM_008770834.1 | XP_008769056.1 | 5, 6 | 905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp1| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031323.1 | NP_112613.1 | 5, 6 | 871 | ||
| XM_006252285.1 | XP_006252347.1 | 2, 3 | 344 | |||
| XM_008770834.1 | XP_008769056.1 | 5, 6 | 905 | |||