siRNA Details
| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031323.1 | NP_112613.1 | 14 | 2029 | ||
| XM_006252285.1 | XP_006252347.1 | 11 | 1502 | |||
| XM_008770834.1 | XP_008769056.1 | 14 | 2063 | |||
| GenBank mRNA | AB012139.1 | - | 3 | 447 | ||

Bmp1
Genomic Map