siRNA Details
| Gene Symbol: Bcl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031328.1 | NP_112618.1 | 2 | 409 | ||
| GenBank mRNA | AB016069.1 | - | 2 | 409 | ||
| BC061772.1 | - | 2 | 397 | 5621919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl10| Gene Symbol: Bcl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031328.1 | NP_112618.1 | 2 | 409 | ||
| GenBank mRNA | AB016069.1 | - | 2 | 409 | ||
| BC061772.1 | - | 2 | 397 | 5621919 | ||