siRNA Details
| Gene Symbol: Ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031329.2 | NP_112619.2 | 8, 9 | 1616 | ||
| XM_006231853.2 | XP_006231915.1 | 8, 9 | 1612 | |||
| XM_006231854.2 | XP_006231916.1 | 8, 9 | 1656 | |||
| XM_006231855.2 | XP_006231917.1 | 7, 8 | 1536 | |||
| GenBank mRNA | BC062037.1 | - | 8, 9 | 1616 | 5623744 | |

Ocln
Genomic Map