siRNA Details
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053339.1 | NP_445791.1 | 3 | 208 | ||
XM_008770372.1 | XP_008768594.1 | 3 | 2914 | |||
XM_008770373.1 | XP_008768595.1 | 3 | 2957 | |||
XM_008770374.1 | XP_008768596.1 | 2 | 181 | |||
GenBank mRNA | X95188.1 | - | 3 | 208 |
