siRNA Details
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031769.1 | NP_113957.1 | 4 | 315 | ||
GenBank mRNA | AF020346.1 | - | 4 | 315 | ||
FQ213832.1 | - | 4 | 419 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031769.1 | NP_113957.1 | 4 | 315 | ||
GenBank mRNA | AF020346.1 | - | 4 | 315 | ||
FQ213832.1 | - | 4 | 419 |