siRNA Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031770.3 | NP_113958.1 | 4 | 325 | ||
| XM_008766344.1 | XP_008764566.1 | 5 | 898 | |||
| GenBank mRNA | AF001953.1 | - | 4 | 325 | ||
| AF022086.1 | - | 4 | 297 | |||
| AY552803.1 | - | 4 | 325 | |||
| BC089221.1 | - | 4 | 339 | 7320619 | ||
| DQ120491.1 | - | 4 | 332 | |||
| DQ120492.1 | - | 4 | 332 | |||

Gnb5
Genomic Map