siRNA Details
Gene Symbol: Rabac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031774.1 | NP_113962.1 | 2 | 237 | ||
GenBank mRNA | AF025506.1 | - | 2 | 237 | ||
BC086387.1 | - | 2 | 204 | 7302383 | ||
FQ210487.1 | - | 2 | 230 |
