siRNA Details
Gene Symbol: LOC103689977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233278.2 | XP_006233340.1 | 7 | 776 | ||
XM_006233279.2 | XP_006233341.1 | 6 | 598 | |||
XM_008761476.1 | XP_008759698.1 | 7 | 778 | |||
XM_008761477.1 | XP_008759699.1 | 6 | 601 |
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271984.1 | NP_001258913.1 | 6 | 596 | ||
NM_031775.3 | NP_113963.2 | 7 | 751 | |||
XM_008761527.1 | XP_008759749.1 | 5 | 541 | |||
XM_008761528.1 | XP_008759750.1 | 5 | 541 | |||
GenBank mRNA | AF025670.1 | - | 7 | 769 | ||
BC078785.1 | - | 7 | 716 | 7123179 | ||
DY310578.1 | - | 6 | 583 |
