siRNA Details
| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031788.1 | NP_113976.1 | 3 | 1012 | ||
| XM_006250847.2 | XP_006250909.1 | 1091 | ||||
| XM_006250848.1 | XP_006250910.1 | 4 | 1096 | |||
| XM_006250849.2 | XP_006250911.1 | 4 | 1354 | |||
| GenBank mRNA | AF037199.1 | - | 3 | 1012 | ||

Rest
Genomic Map