siRNA Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031793.1 | NP_113981.1 | 15 | 1817 | ||
XM_006234324.2 | XP_006234386.1 | 15 | 2193 | |||
XM_006234325.2 | XP_006234387.1 | 15 | 2014 | |||
XM_006234326.2 | XP_006234388.1 | 15 | 2137 | |||
XM_006234327.2 | XP_006234389.1 | 14 | 2089 | |||
XM_006234328.2 | XP_006234390.1 | 15 | 2084 | |||
XM_006234330.2 | XP_006234392.1 | 14 | 1910 | |||
XM_006234331.2 | XP_006234393.1 | 14 | 2036 | |||
XM_006234333.2 | XP_006234395.1 | 14 | 2050 | |||
XM_006234334.2 | XP_006234396.1 | 14 | 1871 | |||
XM_006234335.2 | XP_006234397.1 | 14 | 1997 | |||
XM_008761930.1 | XP_008760152.1 | 14 | 2113 | |||
XM_008761931.1 | XP_008760153.1 | 14 | 2074 | |||
GenBank mRNA | AF043642.1 | - | 15 | 1817 | ||
BC085723.1 | - | 14 | 1863 | 7123868 |
