siRNA Details
| Gene Symbol: Slc12a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031798.1 | NP_113986.1 | 12 | 2058 | ||
| XM_006254752.2 | XP_006254814.1 | 12 | 1989 | |||
| GenBank mRNA | AF051561.1 | - | 12 | 2058 | ||
| AF071863.1 | - | 12 | 1341 | |||

Slc12a2
Genomic Map