siRNA Details
| Gene Symbol: Gpr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031806.1 | NP_113994.1 | 2 | 357 | ||
| GenBank mRNA | AF064706.1 | - | 2 | 357 | ||
| U12006.1 | - | 2 | 357 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr6| Gene Symbol: Gpr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031806.1 | NP_113994.1 | 2 | 357 | ||
| GenBank mRNA | AF064706.1 | - | 2 | 357 | ||
| U12006.1 | - | 2 | 357 | |||
Set Membership: |
|