siRNA Details
| Gene Symbol: Defb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031810.1 | NP_113998.1 | 1, 2 | 136 | ||
| GenBank mRNA | AF068860.1 | - | 1, 2 | 112 | ||
| AF093536.1 | - | 1, 2 | 136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb1| Gene Symbol: Defb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031810.1 | NP_113998.1 | 1, 2 | 136 | ||
| GenBank mRNA | AF068860.1 | - | 1, 2 | 112 | ||
| AF093536.1 | - | 1, 2 | 136 | |||