siRNA Details
| Gene Symbol: Omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031817.1 | NP_114005.1 | 1 | 578 | ||
| XM_008771551.1 | XP_008769773.1 | 1 | 1473 | |||
| GenBank mRNA | AF104362.1 | - | 1 | 578 | ||
| Gene Symbol: LOC103690116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771520.1 | XP_008769742.1 | 3395 | |||
| XM_008771521.1 | XP_008769743.1 | 1473 | ||||

Omd
Genomic Map