siRNA Details
Gene Symbol: Dvl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031820.1 | NP_114008.1 | 14, 15 | 1713 | ||
XM_006239598.2 | XP_006239660.1 | 14, 15 | 1952 | |||
XM_008764388.1 | XP_008762610.1 | 13, 14 | 2115 | |||
XM_008764389.1 | XP_008762611.1 | 13, 14 | 2040 | |||
XM_008764391.1 | XP_008762613.1 | 14, 15 | 1816 | |||
GenBank mRNA | AF143545.2 | - | 14, 15 | 1713 | ||
AF143546.2 | - | 14, 15 | 1713 | |||
BC062070.1 | - | 10, 11 | 1975 | |||
BC160920.1 | - | 7, 8 | 787 | 7461890 |
