siRNA Details
| Gene Symbol: Ptms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031975.2 | NP_114181.1 | 2 | 402 | ||
| GenBank mRNA | BC167753.1 | - | 2 | 392 | 7371571 | |
| X16481.1 | - | 2 | 185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptms| Gene Symbol: Ptms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031975.2 | NP_114181.1 | 2 | 402 | ||
| GenBank mRNA | BC167753.1 | - | 2 | 392 | 7371571 | |
| X16481.1 | - | 2 | 185 | |||