siRNA Details
Gene Symbol: Ptms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031975.2 | NP_114181.1 | 2 | 429 | ||
GenBank mRNA | BC167753.1 | - | 2 | 419 | 7371571 | |
M20616.1 | - | 1 | 33 | |||
M33025.1 | - | 1 | 33 | |||
X16481.1 | - | 2 | 212 |
