siRNA Details
| Gene Symbol: Bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139082.3 | NP_620782.1 | 3 | 503 | ||
| GenBank mRNA | AF387513.1 | - | 3 | 503 | ||
| BC070516.1 | - | 3 | 747 | 7100920 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bambi| Gene Symbol: Bambi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139082.3 | NP_620782.1 | 3 | 503 | ||
| GenBank mRNA | AF387513.1 | - | 3 | 503 | ||
| BC070516.1 | - | 3 | 747 | 7100920 | ||