siRNA Details
| Gene Symbol: Sox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053349.1 | NP_445801.1 | 1 | 1248 | ||
| GenBank mRNA | AJ004858.1 | - | 1 | 1248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sox11| Gene Symbol: Sox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053349.1 | NP_445801.1 | 1 | 1248 | ||
| GenBank mRNA | AJ004858.1 | - | 1 | 1248 | ||