siRNA Details
| Gene Symbol: Sox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053349.1 | NP_445801.1 | 1 | 549 | ||
| GenBank mRNA | AJ004858.1 | - | 1 | 549 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009234, BC078643, BC062095, AK146514, AB108675, and others.

Sox11
Genomic Map