siRNA Details
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053350.2 | NP_445802.2 | 16 | 1780 | ||
GenBank mRNA | AF121052.1 | - | 16 | 1685 | ||
BM389725.1 | - | 3 | 361 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053350.2 | NP_445802.2 | 16 | 1780 | ||
GenBank mRNA | AF121052.1 | - | 16 | 1685 | ||
BM389725.1 | - | 3 | 361 |