siRNA Details
| Gene Symbol: Atox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053359.2 | NP_445811.1 | 2 | 118 | ||
| GenBank mRNA | AF127137.1 | - | 2 | 54 | ||
| AF177671.1 | - | 2 | 133 | |||
| BC058458.1 | - | 2 | 142 | 6919265 | ||
| BI281905.1 | - | 3 | 384 | |||
| BM392393.1 | - | 3 | 388 | |||
| FQ220104.1 | - | 2 | 119 | |||

Atox1
Genomic Map