siRNA Details
| Gene Symbol: Efna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168670.1 | NP_001162141.1 | 2 | 647 | ||
| GenBank mRNA | AF131912.1 | - | 1 | 248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efna2| Gene Symbol: Efna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168670.1 | NP_001162141.1 | 2 | 647 | ||
| GenBank mRNA | AF131912.1 | - | 1 | 248 | ||