siRNA Details
| Gene Symbol: Rab6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053366.1 | NP_445818.1 | 4 | 715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab6a| Gene Symbol: Rab6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053366.1 | NP_445818.1 | 4 | 715 | ||