siRNA Details
Gene Symbol: Rab6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053366.1 | NP_445818.1 | 6 | 887 | ||
XM_006229794.1 | XP_006229856.1 | 6 | 972 | |||
GenBank mRNA | AF148210.1 | - | 3 | 240 | ||
FQ212069.1 | - | 6 | 914 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024287, NM_001163663, AB041575, AK160866, AK154012, and others.
Gene Ontology Categories:
