siRNA Details
| Gene Symbol: Kif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053376.1 | NP_445828.1 | 17 | 1819 | ||
| GenBank mRNA | AF155824.1 | - | 4 | 401 | ||
| BC091304.1 | - | 3 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif2a| Gene Symbol: Kif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053376.1 | NP_445828.1 | 17 | 1819 | ||
| GenBank mRNA | AF155824.1 | - | 4 | 401 | ||
| BC091304.1 | - | 3 | 272 | |||